siRNA Details
| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006750.3 | NP_006741.1 | 5 | 1256 | ||
| GenBank mRNA | AF243385.1 | - | 4 | 1045 | ||
| BC048215.1 | - | 5 | 1241 | 5805707 | ||
| U40572.1 | - | 5 | 1256 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNTB2| Gene Symbol: SNTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006750.3 | NP_006741.1 | 5 | 1256 | ||
| GenBank mRNA | AF243385.1 | - | 4 | 1045 | ||
| BC048215.1 | - | 5 | 1241 | 5805707 | ||
| U40572.1 | - | 5 | 1256 | |||