siRNA Details
| Gene Symbol: SRM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003132.2 | NP_003123.2 | 6 | 835 | ||
| XM_017002179.1 | XP_016857668.1 | 6 | 854 | |||
| GenBank mRNA | BC000309.2 | - | 6 | 801 | 2820975 | |
| BC033106.1 | - | 6 | 767 | 5420683 | ||
| M34338.1 | - | 6 | 826 | |||

SRM
Genomic Map