siRNA Details
| Gene Symbol: SRP54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003136.3 | NP_003127.1 | 3, 4 | 531 | ||
| XM_005268024.2 | XP_005268081.1 | 3, 4 | 784 | |||
| XM_011537106.1 | XP_011535408.1 | 3, 4 | 412 | |||
| GenBank mRNA | AK293319.1 | - | 3, 4 | 323 | ||
| AK312853.1 | - | 3, 4 | 476 | |||
| BC000652.2 | - | 3, 4 | 398 | 3343314 | ||
| BC003389.1 | - | 3, 4 | 404 | 3446599 | ||
| U51920.1 | - | 3, 4 | 251 | |||

SRP54
Genomic Map