siRNA Details
| Gene Symbol: SRPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003137.4 | NP_003128.3 | 16 | 2000 | ||
| NR_034069.1 | 16 | 2028 | ||||
| GenBank mRNA | AJ318054.1 | - | 15 | 2398 | ||
| AK292277.1 | - | 16 | 1945 | |||
| AK299591.1 | - | 16 | 1988 | |||
| AK312712.1 | - | 16 | 1892 | |||
| BC038292.1 | - | 16 | 1965 | 4824261 | ||
| BX648663.1 | - | 16 | 1917 | |||
| BX648697.1 | - | 16 | 1918 | |||

SRPK1
Genomic Map