siRNA Details
| Gene Symbol: SSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003145.3 | NP_003136.1 | 3 | 315 | ||
| XM_011509923.2 | XP_011508225.1 | 3 | 340 | |||
| GenBank mRNA | AK222600.1 | - | 3 | 258 | ||
| AK300678.1 | - | 3 | 315 | |||
| AK313821.1 | - | 3 | 315 | |||
| AY251536.1 | - | 3 | 275 | |||
| BC000341.2 | - | 3 | 261 | 2822983 | ||
| BP196192.1 | - | 3 | 315 | |||
| BX649192.1 | - | 3 | 300 | |||
| CR456973.1 | - | 2 | 235 | |||
| D37991.1 | - | 3 | 259 | |||
| X74104.1 | - | 3 | 285 | |||

SSR2
Genomic Map