siRNA Details
| Gene Symbol: SSR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308197.1 | NP_001295126.1 | 1, 2 | 248 | ||
| NM_007107.4 | NP_009038.1 | 1, 2 | 248 | |||
| GenBank mRNA | AF087907.1 | - | 1, 2 | 196 | ||
| AF110647.1 | - | 1, 2 | 193 | |||
| AK304364.1 | - | 1, 2 | 146 | |||
| AK312935.1 | - | 1, 2 | 155 | |||
| BC017203.1 | - | 1, 2 | 194 | 3618589 | ||
| BT006669.1 | - | 1, 2 | 137 | |||
| DA135338.1 | - | 1, 2 | 232 | |||
| DA750309.1 | - | 1, 2 | 232 | |||
| HY035536.1 | - | 1, 2 | 248 | |||

SSR3
Genomic Map