siRNA Details
| Gene Symbol: SSR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204526.1 | NP_001191455.1 | 6 | 538 | ||
| NM_001204527.1 | NP_001191456.1 | 6 | 561 | |||
| NM_006280.2 | NP_006271.1 | 5 | 511 | |||
| NR_037927.1 | 7 | 732 | ||||
| XM_011531186.1 | XP_011529488.1 | 6 | 420 | |||
| XM_011531187.1 | XP_011529489.1 | 6 | 420 | |||
| XM_017029756.1 | XP_016885245.1 | 6 | 1309 | |||
| XM_017029757.1 | XP_016885246.1 | 6 | 1044 | |||
| GenBank mRNA | AK057117.1 | - | 4 | 2094 | ||
| AK094395.1 | - | 5 | 2214 | |||
| AK290493.1 | - | 5 | 417 | |||
| BC003371.1 | - | 5 | 391 | 2900758 | ||
| BE791319.1 | - | 5 | 433 | |||
| BG165852.1 | - | 6 | 545 | |||
| BP202705.1 | - | 6 | 500 | |||
| BT007192.1 | - | 5 | 387 | |||
| CD672615.1 | - | 6 | 603 | |||
| X90583.1 | - | 5 | 417 | |||
| Z69043.1 | - | 5 | 436 | |||

SSR4
Genomic Map