siRNA Details
| Gene Symbol: STAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243835.1 | NP_001230764.1 | 19 | 1932 | ||
| NM_003151.3 | NP_003142.1 | 19 | 1983 | |||
| XM_006712719.3 | XP_006712782.1 | 19 | 1978 | |||
| XM_011511705.2 | XP_011510007.1 | 22 | 2129 | |||
| GenBank mRNA | AB209043.1 | - | 9 | 619 | ||
| AK299976.1 | - | 16 | 1464 | |||
| AK300868.1 | - | 19 | 1736 | |||
| AK301172.1 | - | 17 | 1407 | |||
| BC031212.1 | - | 19 | 1904 | 4830583 | ||
| L78440.1 | - | 19 | 1749 | |||

STAT4
Genomic Map