siRNA Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006948.4 | NP_008879.3 | 2 | 130 | ||
| GenBank mRNA | AK128540.1 | - | 2 | 88 | ||
| AK223282.1 | - | 2 | 88 | |||
| AK299004.1 | - | 2 | 79 | |||
| AK312396.1 | - | 2 | 85 | |||
| BC036370.2 | - | 2 | 92 | 4837136 | ||
| BI548162.1 | - | 2 | 156 | |||
| U04735.1 | - | 2 | 97 | |||

HSPA13
Genomic Map