siRNA Details
| Gene Symbol: SULT1E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005420.2 | NP_005411.1 | 3 | 300 | ||
| XM_011532210.2 | XP_011530512.1 | 3 | 299 | |||
| GenBank mRNA | AK310845.1 | - | 3 | 293 | ||
| AK312236.1 | - | 2 | 199 | |||
| BC027956.1 | - | 3 | 300 | 5210178 | ||
| CR407621.1 | - | 2 | 187 | |||
| S77383.1 | - | 3 | 223 | |||
| U08098.1 | - | 3 | 293 | |||
| Y11195.1 | - | 3 | 249 | |||

SULT1E1
Genomic Map