siRNA Details
| Gene Symbol: STIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277961.1 | NP_001264890.1 | 6 | 1261 | ||
| NM_001277962.1 | NP_001264891.1 | 6 | 1261 | |||
| NM_003156.3 | NP_003147.2 | 6 | 1261 | |||
| GenBank mRNA | AK310510.1 | - | 3 | 387 | ||
| AK314928.1 | - | 6 | 782 | |||
| BC021300.2 | - | 6 | 1253 | 4899542 | ||
| HQ317451.1 | - | 6 | 1260 | |||
| JX014264.1 | - | 6 | 693 | |||
| U52426.1 | - | 6 | 1258 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009287, U47323, BC021644, AK200235, AK041944, and others.

STIM1
Genomic Map