siRNA Details
| Gene Symbol: CDKL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037343.1 | NP_001032420.1 | 14, 15 | 2293 | ||
| NM_001323289.1 | NP_001310218.1 | 12, 13 | 2206 | |||
| NM_003159.2 | NP_003150.1 | 13, 14 | 2297 | |||
| GenBank mRNA | AK307444.1 | - | 13, 14 | 2294 | ||
| AY217744.1 | - | 13, 14 | 2265 | |||
| HQ171445.1 | - | 12, 13 | 2044 | |||
| X89059.1 | - | 2, 3 | 1030 | |||
| Y15057.1 | - | 13, 14 | 2265 | |||

CDKL5
Genomic Map