siRNA Details
| Gene Symbol: STXBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007269.2 | NP_009200.2 | 7 | 536 | ||
| GenBank mRNA | AF032922.1 | - | 7 | 518 | ||
| AK223170.1 | - | 7 | 515 | |||
| AK290134.1 | - | 7 | 520 | |||
| AK291322.1 | - | 7 | 516 | |||
| AK312910.1 | - | 7 | 513 | |||
| BC038099.2 | - | 7 | 475 | 4826366 | ||
| BC047764.1 | - | 7 | 511 | 5585614 | ||
| D63506.1 | - | 7 | 512 | |||

STXBP3
Genomic Map