siRNA Details
| Gene Symbol: SULT1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001056.3 | NP_001047.1 | 6 | 974 | ||
| NM_176825.2 | NP_789795.1 | 7 | 1007 | |||
| XM_005264013.3 | XP_005264070.1 | 6 | 795 | |||
| GenBank mRNA | AB008164.1 | - | 6 | 644 | ||
| AF026303.1 | - | 6 | 606 | |||
| AF186251.1 | - | 6 | 839 | |||
| AF186252.1 | - | 6 | 974 | |||
| AF186253.1 | - | 6 | 690 | |||
| AF186254.1 | - | 7 | 872 | |||
| AF186255.1 | - | 7 | 1007 | |||
| AF186256.1 | - | 7 | 723 | |||
| AK297086.1 | - | 6 | 780 | |||
| AK298306.1 | - | 6 | 645 | |||
| AK313193.1 | - | 6 | 738 | |||
| AL832145.1 | - | 2 | 5748 | |||
| BC005353.1 | - | 6 | 591 | 4051264 | ||
| BT006951.1 | - | 5 | 521 | |||
| U66036.1 | - | 6 | 697 | |||

SULT1C2
Genomic Map