siRNA Details
| Gene Symbol: SURF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001280787.1 | NP_001267716.1 | 5, 6 | 566 | ||
| NM_003172.3 | NP_003163.1 | 6, 7 | 618 | |||
| XM_011518942.1 | XP_011517244.1 | 5, 6 | 668 | |||
| GenBank mRNA | AK291122.1 | - | 6, 7 | 601 | ||
| BC028314.1 | - | 6, 7 | 606 | 5190921 | ||
| BC071658.1 | - | 6, 7 | 623 | 5539506 | ||
| KU178341.1 | - | 6, 7 | 586 | |||
| KU178342.1 | - | 5, 6 | 534 | |||
| KU178343.1 | - | 4, 5 | 400 | |||
| Z35093.1 | - | 6, 7 | 600 | |||

SURF1
Genomic Map