siRNA Details
| Gene Symbol: TCF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003200.3 | NP_003191.1 | 18 | 1764 | ||
| XM_006722852.3 | XP_006722915.1 | 18 | 1939 | |||
| XM_006722853.3 | XP_006722916.1 | 18 | 1939 | |||
| XM_011528215.2 | XP_011526517.1 | 17 | 3237 | |||
| XM_011528216.2 | XP_011526518.1 | 18 | 1938 | |||
| XM_011528217.2 | XP_011526519.1 | 18 | 1941 | |||
| XM_011528219.2 | XP_011526521.1 | 18 | 1936 | |||
| XM_011528223.2 | XP_011526525.1 | 18 | 1847 | |||
| XM_017027177.1 | XP_016882666.1 | 18 | 1938 | |||
| XM_017027180.1 | XP_016882669.1 | 15 | 1865 | |||
| XM_017027181.1 | XP_016882670.1 | 18 | 1850 | |||
| XM_017027183.1 | XP_016882672.1 | 18 | 2022 | |||
| XM_017027184.1 | XP_016882673.1 | 18 | 1942 | |||
| GenBank mRNA | BC011665.2 | - | 15 | 1508 | ||
| BC110580.1 | - | 14 | 1735 | 40007779 | ||
| M24405.1 | - | 9 | 1053 | |||
| M31222.1 | - | 1764 | ||||
| M31523.1 | - | 17 | 1725 | |||

TCF3
Genomic Map