siRNA Details
| Gene Symbol: TCTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022171.2 | NP_071503.1 | 2 | 453 | ||
| GenBank mRNA | AK000824.1 | - | 2 | 270 | ||
| AK311839.1 | - | 2 | 236 | |||
| BC005157.1 | - | 2 | 419 | 2959904 | ||
| CR457411.1 | - | 2 | 232 | |||
| L41143.1 | - | 2 | 453 | |||

TCTA
Genomic Map