siRNA Details
| Gene Symbol: TCTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022171.2 | NP_071503.1 | 2, 3 | 491 | ||
| GenBank mRNA | AK000824.1 | - | 2, 3 | 308 | ||
| AK311839.1 | - | 2, 3 | 274 | |||
| BC005157.1 | - | 2, 3 | 457 | 2959904 | ||
| CR457411.1 | - | 2, 3 | 270 | |||
| L41143.1 | - | 2, 3 | 491 | |||

TCTA
Genomic Map