siRNA Details
Gene Symbol: TEAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021961.5 | NP_068780.2 | 7, 8 | 1129 | ||
GenBank mRNA | BC115398.1 | - | 7, 8 | 634 | 40068609 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TEAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021961.5 | NP_068780.2 | 7, 8 | 1129 | ||
GenBank mRNA | BC115398.1 | - | 7, 8 | 634 | 40068609 |