siRNA Details
| Gene Symbol: TEAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003213.3 | NP_003204.2 | 10 | 1136 | ||
| NM_201441.2 | NP_958849.1 | 9 | 1007 | |||
| NM_201443.2 | NP_958851.1 | 8 | 1077 | |||
| GenBank mRNA | AK222947.1 | - | 10 | 1023 | ||
| AY101179.1 | - | 9 | 989 | |||
| BC015497.1 | - | 10 | 1034 | 3913870 | ||
| BX640921.1 | - | 8 | 743 | |||
| KU178354.1 | - | 7 | 625 | |||
| KU178355.1 | - | 4 | 396 | |||
| KU178356.1 | - | 3 | 256 | |||
| U63824.1 | - | 10 | 1007 | |||
| X94438.1 | - | 8 | 823 | |||

TEAD4
Genomic Map