siRNA Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006285.2 | NP_006276.2 | 4, 5 | 880 | ||
| GenBank mRNA | AF507907.1 | - | 3, 4 | 293 | ||
| AK298907.1 | - | 4, 5 | 418 | |||
| BC037561.1 | - | 4, 5 | 475 | |||
| BC038448.1 | - | 4, 5 | 854 | 5170888 | ||
| BC067130.1 | - | 4, 5 | 891 | 6153652 | ||
| D50863.1 | - | 4, 5 | 816 | |||

TESK1
Genomic Map