siRNA Details
| Gene Symbol: TFCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173452.1 | NP_001166923.1 | 4 | 1162 | ||
| NM_001173453.1 | NP_001166924.1 | 4 | 1162 | |||
| NM_005653.4 | NP_005644.2 | 4 | 1162 | |||
| GenBank mRNA | AK291264.1 | - | 4 | 813 | ||
| AK308087.1 | - | 4 | 513 | |||
| BC003634.2 | - | 4 | 900 | 3529119 | ||
| M84810.1 | - | 4 | 804 | |||
| U03494.1 | - | 4 | 1132 | |||
| U03495.1 | - | 5 | 949 | |||

TFCP2
Genomic Map