siRNA Details
| Gene Symbol: TFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001206855.1 | NP_001193784.1 | 13 | 1419 | ||
| NM_003227.3 | NP_003218.2 | 16 | 1839 | |||
| XM_005250553.4 | XP_005250610.1 | 17 | 2145 | |||
| XM_017012573.1 | XP_016868062.1 | 17 | 1947 | |||
| GenBank mRNA | AF067864.1 | - | 16 | 1825 | ||
| AK000421.1 | - | 12 | 1151 | |||
| BC142630.1 | - | 16 | 1825 | 40147690 | ||
| BC150192.1 | - | 12 | 1718 | |||

TFR2
Genomic Map