siRNA Details
| Gene Symbol: TGM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003241.3 | NP_003232.2 | 11 | 1536 | ||
| XM_011534042.1 | XP_011532344.1 | 12 | 1685 | |||
| GenBank mRNA | AK300523.1 | - | 11 | 1405 | ||
| AK312936.1 | - | 11 | 1534 | |||
| BC007003.1 | - | 11 | 1531 | 3950865 | ||
| L34840.1 | - | 11 | 1507 | |||
| U31905.1 | - | 11 | 1507 | |||

TGM4
Genomic Map