siRNA Details
| Gene Symbol: TGM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003241.3 | NP_003232.2 | 12 | 1723 | ||
| XM_011534042.1 | XP_011532344.1 | 13 | 1872 | |||
| GenBank mRNA | AK300523.1 | - | 12 | 1592 | ||
| AK312936.1 | - | 12 | 1721 | |||
| BC007003.1 | - | 12 | 1718 | 3950865 | ||
| L34840.1 | - | 12 | 1694 | |||
| U31905.1 | - | 12 | 1694 | |||

TGM4
Genomic Map