siRNA Details
| Gene Symbol: THBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003247.3 | NP_003238.2 | 22, 23 | 3822 | ||
| GenBank mRNA | AI130835.1 | - | 1, 2 | 436 | ||
| AK292429.1 | - | 22, 23 | 3779 | |||
| BC146676.1 | - | 21, 22 | 3629 | 40148868 | ||
| BC150175.1 | - | 21, 22 | 3633 | 40147958 | ||
| BX641023.1 | - | 8, 9 | 1277 | |||
| L12350.1 | - | 22, 23 | 3741 | |||
| M81339.1 | - | 11, 12 | 1826 | |||

THBS2
Genomic Map