siRNA Details
| Gene Symbol: THBS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003247.3 | NP_003238.2 | 4 | 696 | ||
| GenBank mRNA | AK292429.1 | - | 4 | 653 | ||
| BC146676.1 | - | 3 | 503 | 40148868 | ||
| BC150175.1 | - | 3 | 507 | 40147958 | ||
| BX390187.2 | - | 2 | 368 | |||
| L12350.1 | - | 4 | 615 | |||

THBS2
Genomic Map