siRNA Details
| Gene Symbol: THBS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306212.1 | NP_001293141.1 | 23 | 2941 | ||
| NM_001306213.1 | NP_001293142.1 | 23 | 2993 | |||
| NM_001306214.1 | NP_001293143.1 | 22 | 2935 | |||
| NM_003248.5 | NP_003239.2 | 22 | 3107 | |||
| GenBank mRNA | AK313587.1 | - | 22 | 2923 | ||
| BC050456.1 | - | 23 | 3191 | 6191808 | ||
| Z19585.1 | - | 22 | 2895 | |||

THBS4
Genomic Map