siRNA Details
| Gene Symbol: TIMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003254.2 | NP_003245.1 | 3, 4 | 386 | ||
| XM_017029766.1 | XP_016885255.1 | 3, 4 | 340 | |||
| GenBank mRNA | A10416.1 | - | 3, 4 | 256 | ||
| AK074854.1 | - | 3, 4 | 364 | |||
| AK310206.1 | - | 3, 4 | 238 | |||
| AK311937.1 | - | 3, 4 | 238 | |||
| BC000866.1 | - | 3, 4 | 339 | 3460153 | ||
| BC007097.1 | - | 3, 4 | 287 | |||
| BQ181804.1 | - | 3, 4 | 542 | |||
| BU857950.1 | - | 433 | ||||
| CR407638.1 | - | 2, 3 | 194 | |||
| CR541982.1 | - | 2, 3 | 194 | |||
| M12670.1 | - | 3, 4 | 242 | |||
| M59906.1 | - | 3, 4 | 237 | |||
| S68252.1 | - | 2, 3 | 197 | |||
| X02598.1 | - | 3, 4 | 235 | |||
| X03124.1 | - | 3, 4 | 256 | |||

TIMP1
Genomic Map