siRNA Details
| Gene Symbol: TIMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003256.3 | NP_003247.1 | 3 | 774 | ||
| GenBank mRNA | AK313018.1 | - | 3 | 335 | ||
| BC010553.1 | - | 3 | 774 | 4154090 | ||
| BT019627.1 | - | 3 | 263 | |||
| U76456.1 | - | 3 | 322 | |||

TIMP4
Genomic Map