siRNA Details
| Gene Symbol: SEC62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003262.3 | NP_003253.1 | 8 | 1008 | ||
| XM_011513114.2 | XP_011511416.1 | 9 | 1085 | |||
| GenBank mRNA | AK294504.1 | - | 3 | 172 | ||
| BC012035.1 | - | 8 | 992 | 4706586 | ||
| BX648539.1 | - | 8 | 992 | |||
| D87127.1 | - | 8 | 989 | |||
| U93239.1 | - | 8 | 989 | |||

SEC62
Genomic Map