siRNA Details
| Gene Symbol: TSPAN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004615.3 | NP_004606.2 | 3 | 385 | ||
| GenBank mRNA | AB062057.1 | - | 3 | 386 | ||
| AB102665.1 | - | 3 | 301 | |||
| AK293178.1 | - | 4 | 370 | |||
| AK293534.1 | - | 5 | 539 | |||
| AK293552.1 | - | 4 | 450 | |||
| AK312343.1 | - | 3 | 319 | |||
| BC018036.1 | - | 3 | 344 | 4801388 | ||
| D10653.1 | - | 3 | 325 | |||
| D29808.1 | - | 3 | 311 | |||
| L10373.1 | - | 3 | 369 | |||

TSPAN7
Genomic Map