siRNA Details
Gene Symbol: TP53BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031685.2 | NP_001026855.2 | 8 | 1253 | ||
NM_005426.2 | NP_005417.1 | 9 | 1385 | |||
XM_011544268.1 | XP_011542570.1 | 7 | 1114 | |||
XM_011544269.2 | XP_011542571.1 | 5 | 772 | |||
XM_017002220.1 | XP_016857709.1 | 9 | 1394 | |||
GenBank mRNA | AJ318888.2 | - | 8 | 1198 | ||
AK294432.1 | - | 8 | 1105 | |||
AK295378.1 | - | 1 | 31 | |||
BC040247.1 | - | 6 | 729 | 4454266 | ||
BC046150.1 | - | 8 | 1196 | |||
BC058918.1 | - | 8 | 1211 | 4537206 | ||
U58334.1 | - | 9 | 1330 |
