siRNA Details
| Gene Symbol: TUB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003320.4 | NP_003311.2 | 12 | 1720 | ||
| NM_177972.2 | NP_813977.1 | 11 | 1463 | |||
| XM_005253109.3 | XP_005253166.1 | 11 | 1455 | |||
| XM_011520344.2 | XP_011518646.1 | 11 | 1510 | |||
| GenBank mRNA | AK293528.1 | - | 11 | 1391 | ||
| BC075031.2 | - | 12 | 1512 | 30915625 | ||
| BC075032.2 | - | 12 | 1512 | 30915418 | ||
| U54644.1 | - | 11 | 1466 | |||
| U82467.1 | - | 12 | 1720 | |||

TUB
Genomic Map