siRNA Details
| Gene Symbol: TYRO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006293.3 | NP_006284.2 | 8 | 1259 | ||
| XM_017022542.1 | XP_016878031.1 | 8 | 1224 | |||
| XM_017022543.1 | XP_016878032.1 | 8 | 1060 | |||
| GenBank mRNA | BC034960.1 | - | 8 | 1041 | ||
| BC049368.1 | - | 8 | 1033 | 4821587 | ||
| BC051756.1 | - | 8 | 1038 | 6095596 | ||
| D17517.1 | - | 8 | 1247 | |||
| U02566.1 | - | 5 | 470 | |||
| U05682.1 | - | 8 | 1029 | |||

TYRO3
Genomic Map