siRNA Details
| Gene Symbol: UFD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035247.2 | NP_001030324.2 | 4 | 331 | ||
| NM_005659.6 | NP_005650.2 | 4 | 331 | |||
| XM_011530361.2 | XP_011528663.1 | 4 | 457 | |||
| GenBank mRNA | AF141201.1 | - | 3 | 201 | ||
| AJ239058.1 | - | 3 | 201 | |||
| AK225877.1 | - | 4 | 288 | |||
| AK304734.1 | - | 4 | 301 | |||
| AK308786.1 | - | 4 | 275 | |||
| AK310950.1 | - | 4 | 283 | |||
| AK315998.1 | - | 4 | 282 | |||
| AY101594.1 | - | 4 | 258 | |||
| BC001049.2 | - | 4 | 278 | 3507963 | ||
| BC005087.1 | - | 4 | 279 | 3830774 | ||
| BM761958.1 | - | 3 | 270 | |||
| BX648483.1 | - | 4 | 263 | |||
| CB128364.1 | - | 4 | 331 | |||
| CR456607.1 | - | 4 | 215 | |||
| DB496066.1 | - | 4 | 315 | |||
| U64444.1 | - | 4 | 279 | |||

UFD1L
Genomic Map