siRNA Details
| Gene Symbol: XPNPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003399.5 | NP_003390.4 | 20 | 2009 | ||
| GenBank mRNA | BC126174.1 | - | 20 | 1926 | 8991890 | |
| BC143901.1 | - | 20 | 1926 | 9052419 | ||
| BQ224207.1 | - | 7 | 450 | |||
| U90724.2 | - | 20 | 2029 | |||

XPNPEP2
Genomic Map