siRNA Details
| Gene Symbol: ZNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282795.1 | NP_001269724.1 | 5 | 816 | ||
| NM_001282797.1 | NP_001269726.1 | 4 | 656 | |||
| NM_003416.3 | NP_003407.1 | 5 | 783 | |||
| XM_006716654.2 | XP_006716717.1 | 3 | 547 | |||
| XM_006716656.3 | XP_006716719.1 | 3 | 536 | |||
| XM_011517291.2 | XP_011515593.1 | 5 | 830 | |||
| XM_011517292.2 | XP_011515594.1 | 5 | 1164 | |||
| XM_011517293.2 | XP_011515595.1 | 5 | 1163 | |||
| XM_011517294.2 | XP_011515596.1 | 5 | 942 | |||
| XM_011517295.2 | XP_011515597.1 | 5 | 723 | |||
| XM_011517296.2 | XP_011515598.1 | 5 | 734 | |||
| XM_011517297.2 | XP_011515599.1 | 3 | 583 | |||
| XM_017013813.1 | XP_016869302.1 | 5 | 827 | |||
| XM_017013814.1 | XP_016869303.1 | 4 | 1129 | |||
| XM_017013815.1 | XP_016869304.1 | 4 | 1126 | |||
| GenBank mRNA | AB209619.1 | - | 2 | 4172 | ||
| AK300001.1 | - | 5 | 751 | |||
| AK301561.1 | - | 4 | 617 | |||
| AK308002.1 | - | 5 | 775 | |||
| AK313745.1 | - | 5 | 723 | |||
| BC058923.1 | - | 5 | 727 | 4516166 | ||
| BX647998.1 | - | 5 | 742 | |||
| M29580.1 | - | 5 | 884 | |||

ZNF7
Genomic Map