siRNA Details
| Gene Symbol: ZNF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015394.4 | NP_056209.2 | 4, 5 | 445 | ||
| GenBank mRNA | AK314784.1 | - | 4, 5 | 706 | ||
| AL110188.1 | - | 2, 3 | 162 | |||
| BC024182.2 | - | 4, 5 | 446 | 4823159 | ||
| X52332.1 | - | 3, 4 | 329 | |||

ZNF10
Genomic Map