siRNA Details
| Gene Symbol: ZNF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021030.2 | NP_066358.2 | 4 | 578 | ||
| GenBank mRNA | AB021644.1 | - | 4 | 528 | ||
| AK074832.1 | - | 4 | 563 | |||
| BC136323.1 | - | 4 | 465 | 9020310 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF14| Gene Symbol: ZNF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021030.2 | NP_066358.2 | 4 | 578 | ||
| GenBank mRNA | AB021644.1 | - | 4 | 528 | ||
| AK074832.1 | - | 4 | 563 | |||
| BC136323.1 | - | 4 | 465 | 9020310 | ||