siRNA Details
| Gene Symbol: ZNF17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006959.2 | NP_008890.2 | 3 | 1596 | ||
| XM_017027205.1 | XP_016882694.1 | 4 | 3546 | |||
| GenBank mRNA | AB075827.1 | - | 1 | 1203 | ||
| AK097123.1 | - | 1 | 882 | |||
| AK098183.1 | - | 3 | 1578 | |||
| AK127948.1 | - | 4 | 1635 | |||
| AK128471.1 | - | 1 | 1801 | |||
| X52341.1 | - | 1 | 42 | |||

ZNF17
Genomic Map