siRNA Details
| Gene Symbol: ZNF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006961.3 | NP_008892.2 | 5 | 443 | ||
| GenBank mRNA | AK055567.1 | - | 4 | 324 | ||
| AK057330.1 | - | 4 | 618 | |||
| AK292260.1 | - | 5 | 641 | |||
| AK311003.1 | - | 3 | 766 | |||
| BC047091.1 | - | 5 | 430 | 5270853 | ||

ZNF19
Genomic Map