siRNA Details
| Gene Symbol: ZNF32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005368.2 | NP_001005368.1 | 2, 3 | 342 | ||
| NM_006973.2 | NP_008904.1 | 2, 3 | 204 | |||
| XM_017016616.1 | XP_016872105.1 | 1, 2 | 3275 | |||
| GenBank mRNA | AI871265.1 | - | 2, 3 | 247 | ||
| AK312132.1 | - | 2, 3 | 201 | |||
| BC022842.1 | - | 1, 2 | 139 | 5014207 | ||
| BG477527.1 | - | 2, 3 | 206 | |||
| U69645.1 | - | 2, 3 | 180 | |||

ZNF32
Genomic Map