siRNA Details
| Gene Symbol: ZNF33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278170.1 | NP_001265099.1 | 4 | 1862 | ||
| NM_001278171.1 | NP_001265100.1 | 6 | 2041 | |||
| NM_001278173.1 | NP_001265102.1 | 6 | 1997 | |||
| NM_001278174.1 | NP_001265103.1 | 6 | 1993 | |||
| NM_001278175.1 | NP_001265104.1 | 6 | 1982 | |||
| NM_001278176.1 | NP_001265105.1 | 6 | 1998 | |||
| NM_001278177.1 | NP_001265106.1 | 5 | 1914 | |||
| NM_001278178.1 | NP_001265107.1 | 5 | 1923 | |||
| NM_001278179.1 | NP_001265108.1 | 6 | 2038 | |||
| NM_001324175.1 | NP_001311104.1 | 6 | 1990 | |||
| NM_001324176.1 | NP_001311105.1 | 5 | 1920 | |||
| NM_001324177.1 | NP_001311106.1 | 6 | 1998 | |||
| NM_001324178.1 | NP_001311107.1 | 4 | 1865 | |||
| NM_006954.1 | NP_008885.1 | 5 | 1918 | |||
| NM_006974.2 | NP_008905.1 | 5 | 1915 | |||
| XM_011519650.2 | XP_011517952.1 | 6 | 1987 | |||
| XM_011519651.2 | XP_011517953.1 | 6 | 1984 | |||
| XM_011519653.2 | XP_011517955.1 | 4 | 1800 | |||
| XM_011519654.2 | XP_011517956.1 | 4 | 1797 | |||
| XM_017016617.1 | XP_016872106.1 | 5 | 1881 | |||
| GenBank mRNA | AK292844.1 | - | 5 | 1899 | ||
| AK303445.1 | - | 4 | 1857 | |||
| AY184389.1 | - | 3 | 1737 | |||
| BC126119.1 | - | 3 | 1790 | 8991835 | ||
| D31763.1 | - | 5 | 1852 | |||

ZNF33A
Genomic Map