siRNA Details
| Gene Symbol: ZNF134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003435.4 | NP_003426.3 | 2, 3 | 387 | ||
| GenBank mRNA | AK291763.1 | - | 2, 3 | 356 | ||
| BC042636.1 | - | 2, 3 | 346 | 4826076 | ||
| BC112294.1 | - | 2, 3 | 147 | 8327762 | ||
| BC113410.1 | - | 2, 3 | 144 | 8322462 | ||
| DA098572.1 | - | 2, 3 | 387 | |||
| U09412.1 | - | 2, 3 | 330 | |||

ZNF134
Genomic Map