siRNA Details
| Gene Symbol: ZNF157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003446.3 | NP_003437.2 | 4 | 1096 | ||
| GenBank mRNA | BC075003.2 | - | 4 | 1068 | 30915612 | |
| CO245585.1 | - | 622 | ||||
| U28687.1 | - | 4 | 1113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF157| Gene Symbol: ZNF157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003446.3 | NP_003437.2 | 4 | 1096 | ||
| GenBank mRNA | BC075003.2 | - | 4 | 1068 | 30915612 | |
| CO245585.1 | - | 622 | ||||
| U28687.1 | - | 4 | 1113 | |||