siRNA Details
| Gene Symbol: VEZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007146.2 | NP_009077.2 | 2 | 307 | ||
| XM_005257643.1 | XP_005257700.1 | 2 | 286 | |||
| XM_005257644.4 | XP_005257701.1 | 3 | 442 | |||
| XM_017025017.1 | XP_016880506.1 | 3 | 511 | |||
| XM_017025018.1 | XP_016880507.1 | 3 | 363 | |||
| GenBank mRNA | BG718897.1 | - | 3 | 475 | ||
| D28118.1 | - | 2 | 205 | |||

VEZF1
Genomic Map