siRNA Details
| Gene Symbol: ZNF180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278508.2 | NP_001265437.2 | 4 | 843 | ||
| NM_001278509.2 | NP_001265438.2 | 5 | 846 | |||
| NM_001288759.2 | NP_001275688.2 | 5 | 923 | |||
| NM_001288760.2 | NP_001275689.1 | 7 | 1091 | |||
| NM_001288761.2 | NP_001275690.1 | 4 | 719 | |||
| NM_001288762.2 | NP_001275691.1 | 4 | 796 | |||
| NM_001291633.1 | NP_001278562.1 | 5 | 841 | |||
| NM_013256.5 | NP_037388.3 | 5 | 918 | |||
| XM_011527280.2 | XP_011525582.1 | 5 | 3251 | |||
| XM_017027250.1 | XP_016882739.1 | 5 | 992 | |||
| XM_017027251.1 | XP_016882740.1 | 6 | 1061 | |||
| XM_017027252.1 | XP_016882741.1 | 6 | 967 | |||
| XM_017027253.1 | XP_016882742.1 | 6 | 1036 | |||
| XM_017027254.1 | XP_016882743.1 | 4 | 602 | |||
| XM_017027255.1 | XP_016882744.1 | 6 | 1054 | |||
| XM_017027256.1 | XP_016882745.1 | 6 | 1106 | |||
| GenBank mRNA | AF192913.1 | - | 6 | 900 | ||
| AK024047.1 | - | 5 | 843 | |||
| AK122688.1 | - | 4 | 840 | |||
| AK315193.1 | - | 5 | 917 | |||
| BC033642.1 | - | 5 | 910 | 5527054 | ||
| BC113015.1 | - | 5 | 831 | 40034649 | ||

ZNF180
Genomic Map