siRNA Details
| Gene Symbol: ZNF184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318891.1 | NP_001305820.1 | 6 | 942 | ||
| NM_001318892.1 | NP_001305821.1 | 6 | 790 | |||
| NM_001318893.1 | NP_001305822.1 | 6 | 971 | |||
| NM_007149.2 | NP_009080.2 | 6 | 698 | |||
| XM_011514861.2 | XP_011513163.1 | 6 | 724 | |||
| GenBank mRNA | AK123011.1 | - | 5 | 1224 | ||
| AK131264.1 | - | 6 | 950 | |||
| AK312804.1 | - | 6 | 939 | |||
| BC022992.1 | - | 6 | 680 | 4997766 | ||
| U66561.1 | - | 3 | 338 | |||

ZNF184
Genomic Map