siRNA Details
| Gene Symbol: ZNF215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013250.2 | NP_037382.2 | 3 | 602 | ||
| XM_006718311.2 | XP_006718374.1 | 3 | 642 | |||
| GenBank mRNA | AF056618.1 | - | 3 | 602 | ||
| AK298365.1 | - | 3 | 562 | |||
| AK302639.1 | - | 3 | 645 | |||
| BC014538.1 | - | 1 | 104 | 3827937 | ||
| CN483744.1 | - | 3 | 562 | |||

ZNF215
Genomic Map